ENA292596623 Enamine

3-amino-N-(3-chloro-2-methylphenyl)propanamide

  • MDL number MFCD09726265
  • Cas Number : 938517-08-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-62422

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-62422
Availability details for
Quantity
  • Availability
  • Add to Cart