ENA292596628 Enamine

2-(4-bromobenzenesulfonyl)-2-methylpropanenitrile

  • MDL number MFCD12816678
  • Cas Number : 1178187-96-6
  • Molecular Weight : 288.16

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-62428

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-62428
Availability details for
Quantity
  • Availability
  • Add to Cart