ENA292596629 Enamine

1-(4-aminopiperidin-1-yl)-2-(2-methoxyphenyl)ethan-1-one

  • MDL number MFCD11164429
  • Cas Number : 1094366-31-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-61506

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-61506
Availability details for
Quantity
  • Availability
  • Add to Cart