ENA292596630 Enamine

4-methyl-2-(thiomorpholine-4-carbonyl)phenol

  • MDL number MFCD11172370
  • Cas Number : 1042634-26-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-62430

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-62430
Availability details for
Quantity
  • Availability
  • Add to Cart