ENA292596633 Enamine

2-(5-bromo-2,3-dioxo-2,3-dihydro-1H-indol-1-yl)-N,N-dimethylacetamide

  • MDL number MFCD11524351
  • Cas Number : 1021023-54-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-62436

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-62436
Availability details for
Quantity
  • Availability
  • Add to Cart