ENA292596634 Enamine

2-(piperazine-1-carbonyl)pyrazine hydrochloride

  • MDL number MFCD09607872
  • Cas Number : 1185312-60-0
  • Molecular Weight : 228.68

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HCl
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-62440

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-62440
Availability details for
Quantity
  • Availability
  • Add to Cart