ENA292596635 Enamine

5-bromo-1-[(1,2,4-oxadiazol-3-yl)methyl]-2,3-dihydro-1H-indole-2,3-dione

  • MDL number MFCD11538287
  • Cas Number : 1042633-64-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form powder
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-62441

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-62441
Availability details for
Quantity
  • Availability
  • Add to Cart