ENA292596636 Enamine

1-methyl-N-[(piperidin-4-yl)methyl]-1H-pyrazole-4-carboxamide

  • MDL number MFCD11168090
  • Cas Number : 1152936-16-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-62442

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-62442
Availability details for
Quantity
  • Availability
  • Add to Cart