ENA292596637 Enamine

1-(pyridin-4-ylmethyl)-1H-1,2,4-triazol-3-amine

  • MDL number MFCD12766717
  • Cas Number : 1182736-00-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 156...158
Purity 95.00
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-62444

Safety Information

msds https://www.enaminestore.com/catalog/EN300-62444
Availability details for
Quantity
  • Availability
  • Add to Cart