ENA292596639 Enamine

2-(cyclobutylformamido)acetic acid

  • MDL number MFCD09048424
  • Cas Number : 604790-72-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-62448

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-62448
Availability details for
Quantity
  • Availability
  • Add to Cart