ENA292596642 Enamine

cyclopropyl(3,4-dichlorophenyl)methanone

  • MDL number MFCD03841239
  • Cas Number : 136906-33-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-62452

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-62452
Availability details for
Quantity
  • Availability
  • Add to Cart