ENA292596649 Enamine

2-(difluoromethoxy)-6-methylaniline

  • MDL number MFCD12170083
  • Cas Number : 139909-66-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-62466

Safety Information

signalword Warning
msds https://www.enaminestore.com/catalog/EN300-62466
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart