ENA292596650 Enamine

2-amino-N-[3-(pyrrolidin-1-yl)phenyl]hexanamide

  • MDL number MFCD16040167
  • Cas Number : 1218166-69-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form powder
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-62467

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-62467
Availability details for
Quantity
  • Availability
  • Add to Cart