ENA292596655 Enamine

[1-(2-methylphenyl)cyclohexyl]methanamine

  • MDL number MFCD11188923
  • Cas Number : 1037131-52-4
  • Molecular Weight : 203.33

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form liquid
Shipping Temperature Normal
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-62479

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-62479
Availability details for
Quantity
  • Availability
  • Add to Cart