ENA292596658 Enamine

2-(cyclopropylmethoxy)benzoic acid

  • MDL number MFCD06208775
  • Cas Number : 136013-76-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-62482

Safety Information

msds https://www.enaminestore.com/catalog/EN300-62482
Availability details for
Quantity
  • Availability
  • Add to Cart