ENA292596662 Enamine

4-(butan-2-ylsulfanyl)-2-(trifluoromethyl)aniline

  • MDL number MFCD11136074
  • Cas Number : 1019539-28-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-61507

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-61507
Availability details for
Quantity
  • Availability
  • Add to Cart