ENA292596663 Enamine

4-(2-methyl-1H-imidazol-1-yl)pyridine-2-carboxylic acid

  • MDL number MFCD11181368
  • Cas Number : 1094369-47-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-62513

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-62513
Availability details for
Quantity
  • Availability
  • Add to Cart