ENA292596664 Enamine

2-methyl-6-(thiomorpholine-4-carbonyl)aniline

  • MDL number MFCD11172028
  • Cas Number : 1042776-39-5
  • Molecular Weight : 236.34

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name 2-methyl-6-(4-thiomorpholinylcarbonyl)aniline
Inchi Code 1S/C12H16N2OS/c1-9-3-2-4-10(11(9)13)12(15)14-5-7-16-8-6-14/h2-4H,5-8,13H2,1H3
Inchi Key RMEPSQNGYOPPQQ-UHFFFAOYSA-N

Description

Purity 95
Storage temp. RT
Physical Form powder
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-62514

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-62514
Availability details for
Quantity
  • Availability
  • Add to Cart