ENA292596666 Enamine

4-(4-chlorophenyl)oxan-4-amine

  • MDL number MFCD09904027
  • Cas Number : 1017450-84-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-62518

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-62518
Availability details for
Quantity
  • Availability
  • Add to Cart