ENA292596667 Enamine

3-[(1-methyl-1H-pyrazol-4-yl)sulfamoyl]thiophene-2-carboxylic acid

  • MDL number MFCD12075670
  • Cas Number : 1153942-75-6
  • Molecular Weight : 287.32

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form powder
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-62520

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-62520
Availability details for
Quantity
  • Availability
  • Add to Cart