ENA292596668 Enamine

2-methyl-6-(thiomorpholine-4-carbonyl)phenol

  • MDL number MFCD11172366
  • Cas Number : 1042803-11-1
  • Molecular Weight : 237.32

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form powder
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-62521

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-62521
Availability details for
Quantity
  • Availability
  • Add to Cart