ENA292596673 Enamine

3-[5-(2-bromo-5-fluorophenyl)-1,3,4-oxadiazol-2-yl]propanoic acid

  • MDL number MFCD12871768
  • Cas Number : 1178951-41-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-62527

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-62527
Availability details for
Quantity
  • Availability
  • Add to Cart