ENA292596679 Enamine

[2-(dimethylamino)ethyl][(1-ethylpyrrolidin-2-yl)methyl]amine

  • MDL number MFCD11153901
  • Cas Number : 1021241-95-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form liquid
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-62539

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-62539
Availability details for
Quantity
  • Availability
  • Add to Cart