ENA292596681 Enamine

1-(4-chlorophenyl)cyclohexan-1-amine

  • MDL number MFCD09904026
  • Cas Number : 1017478-76-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-62541

Safety Information

msds https://www.enaminestore.com/catalog/EN300-62541
Availability details for
Quantity
  • Availability
  • Add to Cart