ENA292596685 Enamine

methyl 1,4-diazepane-1-carboxylate

  • MDL number MFCD11128919
  • Cas Number : 271243-23-3
  • Molecular Weight : 158.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form liquid
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-62545

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-62545
Availability details for
Quantity
  • Availability
  • Add to Cart