ENA292596688 Enamine

2-butyl-2,3-dihydro-1H-isoindol-5-amine

  • MDL number MFCD11655042
  • Cas Number : 1099643-57-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-62550

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-62550
Availability details for
Quantity
  • Availability
  • Add to Cart