ENA292596693 Enamine

1-(2-phenylethyl)-1H-pyrazol-5-amine

  • MDL number MFCD09742444
  • Cas Number : 3524-23-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 67...69
Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-37518

Safety Information

msds https://www.enaminestore.com/catalog/EN300-37518
Availability details for
Quantity
  • Availability
  • Add to Cart