ENA292596695 Enamine

2,3,5,6,7,8-hexahydrocinnolin-3-one

  • MDL number MFCD00828986
  • Cas Number : 5468-36-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 194-196
Purity 95
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-40564

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-40564
Availability details for
Quantity
  • Availability
  • Add to Cart