ENA292596702 Enamine

dimethyl(piperidin-3-ylmethyl)amine dihydrochloride

  • MDL number MFCD13561484
  • Cas Number : 1211495-34-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 261...263
Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-61527

Safety Information

signalword Warning
msds https://www.enaminestore.com/catalog/EN300-61527
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart