ENA292596712 Enamine

4,5-dimethoxy-2-{[(propan-2-yl)carbamoyl]amino}benzoic acid

  • MDL number MFCD01949222
  • Cas Number : 400786-17-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-61533

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-61533
Availability details for
Quantity
  • Availability
  • Add to Cart