ENA292596716 Enamine

N-(3-aminophenyl)furan-3-carboxamide

  • MDL number MFCD09046813
  • Cas Number : 926212-87-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Product Link https://www.enaminestore.com/catalog/EN300-61535

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-61535
Availability details for
Quantity
  • Availability
  • Add to Cart