ENA292596717 Enamine

2-[4-methyl-6-oxo-2-(pyrrolidin-1-yl)-1,6-dihydropyrimidin-5-yl]acetic acid

  • MDL number MFCD13188178
  • Cas Number : 1216122-04-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 262...264
Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-61536

Safety Information

msds https://www.enaminestore.com/catalog/EN300-61536
Availability details for
Quantity
  • Availability
  • Add to Cart