ENA292596719 Enamine

5-{[(2,3-dihydro-1,4-benzodioxin-2-yl)methyl]sulfanyl}-1,3,4-thiadiazol-2-amine

  • MDL number MFCD01626995
  • Cas Number : 309270-86-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-61538

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-61538
Availability details for
Quantity
  • Availability
  • Add to Cart