ENA292596724 Enamine

1-benzyl-N-propylpiperidin-3-amine

  • MDL number MFCD11183086
  • Cas Number : 1094325-69-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Product Link https://www.enaminestore.com/catalog/EN300-45376

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-45376
Availability details for
Quantity
  • Availability
  • Add to Cart