ENA292596729 Enamine

3-methylhexanoic acid

  • MDL number MFCD00870847
  • Cas Number : 3780-58-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-61548

Safety Information

msds https://www.enaminestore.com/catalog/EN300-61548
Availability details for
Quantity
  • Availability
  • Add to Cart