ENA292596731 Enamine

2-{[(5-bromo-2-fluorophenyl)methyl]amino}ethan-1-ol

  • MDL number MFCD10692731
  • Cas Number : 1152522-37-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-61550

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-61550
Availability details for
Quantity
  • Availability
  • Add to Cart