ENA292596734 Enamine

1-(2-fluorophenyl)-4-hydroxy-1H-pyrazole-3-carboxylic acid

  • MDL number MFCD11133585
  • Cas Number : 1152602-04-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-59679

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-59679
Availability details for
Quantity
  • Availability
  • Add to Cart