ENA292596741 Enamine

3-(cyclohexylmethoxy)pyridin-2-amine

  • MDL number MFCD10689932
  • Cas Number : 107229-69-6
  • Molecular Weight : 206.29

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form powder
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-61575

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-61575
Availability details for
Quantity
  • Availability
  • Add to Cart