ENA295068088 Enamine

1-(aminomethyl)cyclopentan-1-amine dihydrochloride

  • MDL number MFCD16547503
  • Cas Number : 123194-04-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-52195

Safety Information

msds https://www.enaminestore.com/catalog/EN300-52195
Availability details for
Quantity
  • Availability
  • Add to Cart