ENA295068097 Enamine

3-(dimethylamino)propanimidamide dihydrochloride

  • MDL number MFCD16547517
  • Cas Number : 1240527-89-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data 2HCl
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-59981

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-59981
Availability details for
Quantity
  • Availability
  • Add to Cart