ENA295068098 Enamine

ethyl 3-[4-(benzyloxy)phenyl]-3-hydroxypropanoate

  • MDL number MFCD16547520
  • Cas Number : 328396-03-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-60661

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-60661
Availability details for
Quantity
  • Availability
  • Add to Cart