ENA295068100 Enamine

1-benzyl-5-(benzylamino)-2-oxo-2,3-dihydro-1H-imidazole-4-carbonitrile

  • MDL number MFCD16547522
  • Cas Number : 1240527-94-9
  • Molecular Weight : 304.35

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form powder
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-60778

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-60778
Availability details for
Quantity
  • Availability
  • Add to Cart