ENA295068101 Enamine

1-(2-fluorophenyl)-5-(trifluoromethyl)-1H-1,2,3-triazole-4-carboxylic acid

  • MDL number MFCD16547523
  • Cas Number : 1240529-31-0
  • Molecular Weight : 275.16

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-60874

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-60874
Availability details for
Quantity
  • Availability
  • Add to Cart