ENA295068105 Enamine

2-(3-aminopropyl)-1$l^{6},2-thiazolidine-1,1-dione hydrochloride

  • MDL number MFCD16547526
  • Cas Number : 1240528-77-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-61494

Safety Information

msds https://www.enaminestore.com/catalog/EN300-61494
Availability details for
Quantity
  • Availability
  • Add to Cart