ENA295068107 Enamine

4-(3-methylpiperidin-1-yl)butanimidamide dihydrochloride

  • MDL number MFCD16547528
  • Cas Number : 1240529-03-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 209...211
Purity 95.00
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-61849

Safety Information

msds https://www.enaminestore.com/catalog/EN300-61849
Availability details for
Quantity
  • Availability
  • Add to Cart