ENA295068109 Enamine

2,2,2-trifluoroethyl N-(prop-2-en-1-yl)carbamate

  • MDL number MFCD16547531
  • Cas Number : 1240527-16-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-61990

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-61990
Availability details for
Quantity
  • Availability
  • Add to Cart