ENA295068120 Enamine

2,2,2-trifluoroethyl N-(5-phenyl-1,2,4-triazin-3-yl)carbamate

  • MDL number MFCD16547541
  • Cas Number : 1240529-55-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-62334

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-62334
Availability details for
Quantity
  • Availability
  • Add to Cart