ENA295068122 Enamine

2,2,2-trifluoroethyl N-{5,7-dimethyl-[1,2,4]triazolo[1,5-a]pyrimidin-2-yl}carbamate

  • MDL number MFCD16547543
  • Cas Number : 1240529-08-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-62341

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-62341
Availability details for
Quantity
  • Availability
  • Add to Cart