ENA295068125 Enamine

8-phenyl-3,4-dihydro-2H-1-benzopyran-4-amine hydrochloride

  • MDL number MFCD16547549
  • Cas Number : 101089-48-9
  • Molecular Weight : 261.75

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HCl
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-62443

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-62443
Availability details for
Quantity
  • Availability
  • Add to Cart