ENA295068126 Enamine

benzyl 2-[(chlorosulfonyl)methyl]pyrrolidine-1-carboxylate

  • MDL number MFCD16547551
  • Cas Number : 1240529-52-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 60...62
Purity 95.00
Storage temp. -20
Shipping Temperature Ice Pack
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-62454

Safety Information

msds https://www.enaminestore.com/catalog/EN300-62454
Availability details for
Quantity
  • Availability
  • Add to Cart